4-Morpholinyl(2-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
Synonyms:
4-(Piperidin-2-ylcarbonyl)morpholine hydrochloride
052423-500MG
052423-2.5G
101899-578EA
364.62
USD
101899-578
101899-580
4-Morpholinyl(2-piperidinyl)methanone hydrochloride
4-Morpholinyl(2-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O₂ MW: 234.73 g/mol |
MDL Number:
MFCD05865124 CAS Number: 690634-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...