1-Piperidinyl(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

Synonyms: 1-(Piperidin-2-ylcarbonyl)piperidine hydrochloride

052422-500MG 052422-2.5G
101899-574EA 249.23 USD
101899-574 101899-576
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
1-Piperidinyl(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MW: 232.76 g/mol
MDL Number: MFCD06407996
CAS Number: 130497-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR