N,N-Diethyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052420-500MG 052420-2.5G
101899-566EA 364.62 USD
101899-566 101899-568
N,N-Diethyl-2-piperidinecarboxamide hydrochloride
N,N-Diethyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD09999010
CAS Number: 130497-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR