2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052419-500MG 052419-2.5G
101899-562EA 364.62 USD
101899-562 101899-564
2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride
2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O
MW: 218.73 g/mol
MDL Number: MFCD06407992
CAS Number: 690634-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR