N,N-Dimethyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052418-500MG 052418-2.5G
101899-558EA 364.62 USD
101899-558 101899-560
N,N-Dimethyl-2-piperidinecarboxamide hydrochloride
N,N-Dimethyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13562175
CAS Number: 383425-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR