N-Benzyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052416-500MG 052416-2.5G
101899-550EA 364.62 USD
101899-550 101899-552
N-Benzyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride
N-Benzyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MDL Number: MFCD13562173
CAS Number: 1236266-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR