3,4-Dihydro-1(2H)-quinolinyl(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052415-500MG 052415-2.5G
101899-546EA 364.62 USD
101899-546 101899-548
3,4-Dihydro-1(2H)-quinolinyl(2-pyrrolidinyl)methanone hydrochloride
3,4-Dihydro-1(2H)-quinolinyl(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD13562172
CAS Number: 1246172-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR