2,3-Dihydro-1H-indol-1-yl-(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052414-500MG 052414-2.5G
101899-542EA 364.62 USD
101899-542 101899-544
2,3-Dihydro-1H-indol-1-yl-(2-pyrrolidinyl)methanone hydrochloride
2,3-Dihydro-1H-indol-1-yl-(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MW: 252.75 g/mol
MDL Number: MFCD13562171
CAS Number: 1246172-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR