N-Cyclohexyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052411-500MG 052411-2.5G
101899-530EA 364.62 USD
101899-530 101899-532
N-Cyclohexyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride
N-Cyclohexyl-N-ethyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O
MDL Number: MFCD13562168
CAS Number: 1236262-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR