N-Benzyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052410-500MG 052410-2.5G
101899-526EA 364.62 USD
101899-526 101899-528
N-Benzyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride
N-Benzyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562167
CAS Number: 1246172-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR