(2-Ethyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052409-500MG 052409-2.5G
101899-522EA 364.62 USD
101899-522 101899-524
(2-Ethyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
(2-Ethyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MW: 246.78 g/mol
MDL Number: MFCD13562166
CAS Number: 1236261-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR