N-Cyclohexyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052408-500MG 052408-2.5G
101899-518EA 364.62 USD
101899-518 101899-520
N-Cyclohexyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride
N-Cyclohexyl-N-methyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MDL Number: MFCD13562165
CAS Number: 1236266-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR