N,N-Dipropyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052407-500MG 052407-2.5G
101899-514EA 364.62 USD
101899-514 101899-516
N,N-Dipropyl-2-pyrrolidinecarboxamide hydrochloride
N,N-Dipropyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562164
CAS Number: 1236263-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR