(4-Hydroxy-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052406-500MG
052406-2.5G
101899-510EA
364.62
USD
101899-510
101899-512
(4-Hydroxy-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O₂ |
MDL Number:
MFCD13562163 CAS Number: 1236256-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...