4-(2-Pyrrolidinylcarbonyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052405-500MG 052405-2.5G
101899-506EA 364.62 USD
101899-506 101899-508
4-(2-Pyrrolidinylcarbonyl)-2-piperazinone hydrochloride
4-(2-Pyrrolidinylcarbonyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClN₃O₂
MDL Number: MFCD13562162
CAS Number: 1236255-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR