(4-Benzyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052403-500MG 052403-2.5G
101899-498EA 364.62 USD
101899-498 101899-500
(4-Benzyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride
(4-Benzyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClN₃O
MDL Number: MFCD13562160
CAS Number: 1236259-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR