(4-Methyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052402-500MG
052402-2.5G
101899-494EA
364.62
USD
101899-494
101899-496
(4-Methyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride
(4-Methyl-1-piperazinyl)(2-pyrrolidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClN₃O |
MDL Number:
MFCD13562159 CAS Number: 1236260-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...