(4-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052401-500MG 052401-2.5G
101899-490EA 364.62 USD
101899-490 101899-492
(4-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MDL Number: MFCD13562158
CAS Number: 1236267-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR