(2-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052399-500MG 052399-2.5G
101899-482EA 364.62 USD
101899-482 101899-484
(2-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride
(2-Methyl-1-piperidinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MW: 232.76 g/mol
MDL Number: MFCD13562156
CAS Number: 1246172-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR