N-Ethyl-N-(2-hydroxyethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052397-500MG
052397-2.5G
101899-474EA
364.62
USD
101899-474
101899-476
N-Ethyl-N-(2-hydroxyethyl)-2-pyrrolidinecarboxamide hydrochloride
N-Ethyl-N-(2-hydroxyethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562154 CAS Number: 1236262-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...