(3-Hydroxy-1-pyrrolidinyl)(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052395-500MG 052395-2.5G
101899-466EA 364.62 USD
101899-466 101899-468
(3-Hydroxy-1-pyrrolidinyl)(2-pyrrolidinyl)methanone hydrochloride
(3-Hydroxy-1-pyrrolidinyl)(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O₂
MW: 220.7 g/mol
MDL Number: MFCD13562152
CAS Number: 1236254-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR