4-Morpholinyl(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052394-500MG 052394-2.5G
101899-462EA 364.62 USD
101899-462 101899-464
4-Morpholinyl(2-pyrrolidinyl)methanone hydrochloride
4-Morpholinyl(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O₂
MDL Number: MFCD11052934
CAS Number: 1096446-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR