1-Piperidinyl(2-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052393-500MG 052393-2.5G
101899-458EA 166.15 USD
101899-458 101899-460
1-Piperidinyl(2-pyrrolidinyl)methanone hydrochloride
1-Piperidinyl(2-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O
MDL Number: MFCD13562151
CAS Number: 1236259-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR