N,N-Diethyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052391-500MG 052391-2.5G
101899-450EA 364.62 USD
101899-450 101899-452
N,N-Diethyl-2-pyrrolidinecarboxamide hydrochloride
N,N-Diethyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MW: 206.72 g/mol
MDL Number: MFCD04545011
CAS Number: 1048649-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR