2-Amino-N-benzyl-N-ethyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052387-500MG 052387-2.5G
101899-434EA 364.62 USD
101899-434 101899-436
2-Amino-N-benzyl-N-ethyl-3-phenylpropanamide hydrochloride
2-Amino-N-benzyl-N-ethyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₃ClN₂O
MDL Number: MFCD13562146
CAS Number: 1236266-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR