2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052386-500MG 052386-2.5G
101899-430EA 364.62 USD
101899-430 101899-432
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-phenyl-1-propanone hydrochloride
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁ClN₂O
MDL Number: MFCD13562145
CAS Number: 1236258-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR