2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052385-500MG 052385-2.5G
101899-426EA 364.62 USD
101899-426 101899-428
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉ClN₂O
MDL Number: MFCD13562144
CAS Number: 1236254-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR