2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052384-500MG 052384-2.5G
101899-422EA 186.15 USD
101899-422 101899-424
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-phenyl-1-propanone hydrochloride
2-Amino-1-[3,4-dihydro-2(1H)-isoquinolinyl]-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁ClN₂O
MDL Number: MFCD13562143
CAS Number: 1236262-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR