2-Amino-N-cyclohexyl-N-ethyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052382-500MG 052382-2.5G
101899-414EA 364.62 USD
101899-414 101899-416
2-Amino-N-cyclohexyl-N-ethyl-3-phenylpropanamide hydrochloride
2-Amino-N-cyclohexyl-N-ethyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇ClN₂O
MDL Number: MFCD13562141
CAS Number: 1236260-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR