2-Amino-N-benzyl-N-methyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052381-500MG 052381-2.5G
101899-410EA 364.62 USD
101899-410 101899-412
2-Amino-N-benzyl-N-methyl-3-phenylpropanamide hydrochloride
2-Amino-N-benzyl-N-methyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₁ClN₂O
MW: 304.82 g/mol
MDL Number: MFCD13562140
CAS Number: 1236259-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR