2-Amino-1-(2-ethyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052380-500MG
052380-2.5G
101899-406EA
364.62
USD
101899-406
101899-408
2-Amino-1-(2-ethyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(2-ethyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₅ClN₂O |
MDL Number:
MFCD13562139 CAS Number: 1236267-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...