2-Amino-1-(4-hydroxy-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052377-500MG
052377-2.5G
101899-394EA
364.62
USD
101899-394
101899-396
2-Amino-1-(4-hydroxy-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(4-hydroxy-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁ClN₂O₂ |
MDL Number:
MFCD13562136 CAS Number: 1236254-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...