4-(2-Amino-3-phenylpropanoyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052376-500MG 052376-2.5G
101899-390EA 364.62 USD
101899-390 101899-392
4-(2-Amino-3-phenylpropanoyl)-2-piperazinone hydrochloride
4-(2-Amino-3-phenylpropanoyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O₂
MDL Number: MFCD13562135
CAS Number: 1236260-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR