2-Amino-1-(4-methyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052373-500MG 052373-2.5G
101899-378EA 364.62 USD
101899-378 101899-380
2-Amino-1-(4-methyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(4-methyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClN₃O
MDL Number: MFCD13562132
CAS Number: 1236254-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR