2-Amino-1-(4-methyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052372-500MG 052372-2.5G
101899-374EA 364.62 USD
101899-374 101899-376
2-Amino-1-(4-methyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(4-methyl-1-piperidinyl)-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O
MDL Number: MFCD13562131
CAS Number: 1246172-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR