2-Amino-N-butyl-N-methyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052367-500MG 052367-2.5G
101899-354EA 364.62 USD
101899-354 101899-356
2-Amino-N-butyl-N-methyl-3-phenylpropanamide hydrochloride
2-Amino-N-butyl-N-methyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₃ClN₂O
MDL Number: MFCD13562126
CAS Number: 1236259-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR