2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-3-phenyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052366-500MG
052366-2.5G
101899-350EA
364.62
USD
101899-350
101899-352
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-3-phenyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂ |
MDL Number:
MFCD13562125 CAS Number: 1236272-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...