2-Amino-1-(4-morpholinyl)-3-phenyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052365-500MG
052365-2.5G
101899-346EA
364.62
USD
101899-346
101899-348
2-Amino-1-(4-morpholinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(4-morpholinyl)-3-phenyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂ |
MDL Number:
MFCD13562124 CAS Number: 68319-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...