2-Amino-N,N-dimethyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052360-500MG 052360-2.5G
101899-326EA 384.62 USD
101899-326 101899-328
2-Amino-N,N-dimethyl-3-phenylpropanamide hydrochloride
2-Amino-N,N-dimethyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD13562120
CAS Number: 1175090-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR