2-Amino-N-2-dimethyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

052359-500MG 052359-2.5G
101899-322EA 364.62 USD
101899-322 101899-324
2-Amino-N-2-dimethyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
2-Amino-N-2-dimethyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD13562119
CAS Number: 1220031-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR