2-Amino-N-benzyl-N-ethyl-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
052358-500MG
052358-2.5G
101899-318EA
364.62
USD
101899-318
101899-320
2-Amino-N-benzyl-N-ethyl-2-methylpropanamide hydrochloride
2-Amino-N-benzyl-N-ethyl-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₁ClN₂O MW: 256.78 g/mol |
MDL Number:
MFCD13562118 CAS Number: 1219972-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...