2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-2-methyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052356-500MG 052356-2.5G
101899-310EA 364.62 USD
101899-310 101899-312
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-2-methyl-1-propanone hydrochloride
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-2-methyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD13562116
CAS Number: 1220034-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR