2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052354-500MG 052354-2.5G
101899-302EA 364.62 USD
101899-302 101899-304
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-1-propanone hydrochloride
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClN₃O₂
MDL Number: MFCD13562114
CAS Number: 1219972-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR