2-Amino-1-(2-ethyl-1-piperidinyl)-2-methyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052351-500MG
052351-2.5G
101899-290EA
364.62
USD
101899-290
101899-292
2-Amino-1-(2-ethyl-1-piperidinyl)-2-methyl-1-propanone hydrochloride
2-Amino-1-(2-ethyl-1-piperidinyl)-2-methyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₃ClN₂O |
MDL Number:
MFCD13562111 CAS Number: 1219972-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...