2-Amino-2-methyl-1-(4-methyl-1-piperazinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052344-500MG
052344-2.5G
101899-262EA
364.62
USD
101899-262
101899-264
2-Amino-2-methyl-1-(4-methyl-1-piperazinyl)-1-propanone hydrochloride
2-Amino-2-methyl-1-(4-methyl-1-piperazinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₉H₂₀ClN₃O |
MDL Number:
MFCD13562104 CAS Number: 1220034-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...