2-Amino-2-methyl-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052341-500MG 052341-2.5G
101899-250EA 364.62 USD
101899-250 101899-252
2-Amino-2-methyl-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride
2-Amino-2-methyl-1-(2-methyl-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562101
CAS Number: 1220036-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR