N,N-Diallyl-2-amino-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052340-500MG 052340-2.5G
101899-246EA 364.62 USD
101899-246 101899-248
N,N-Diallyl-2-amino-2-methylpropanamide hydrochloride
N,N-Diallyl-2-amino-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O
MDL Number: MFCD13562100
CAS Number: 1220031-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR