2-Amino-N-butyl-N-2-dimethylpropanamide hydrochloride

Supplier: Matrix Scientific

052338-500MG 052338-2.5G
101899-238EA 364.62 USD
101899-238 101899-240
2-Amino-N-butyl-N-2-dimethylpropanamide hydrochloride
2-Amino-N-butyl-N-2-dimethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₂₁ClN₂O
MDL Number: MFCD13562098
CAS Number: 1220034-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR