2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-2-methyl-1-propanone hydrochloride
Supplier: Matrix Scientific
052337-500MG
052337-2.5G
101899-234EA
364.62
USD
101899-234
101899-236
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-2-methyl-1-propanone hydrochloride
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-2-methyl-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ MW: 208.69 g/mol |
MDL Number:
MFCD13562097 CAS Number: 1219971-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...