2-Amino-2-methyl-1-(1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052335-500MG 052335-2.5G
101899-226EA 364.62 USD
101899-226 101899-228
2-Amino-2-methyl-1-(1-piperidinyl)-1-propanone hydrochloride
2-Amino-2-methyl-1-(1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562095
CAS Number: 1220034-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR